Assay Details
| Gene Symbol: A930001C03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045989.1 | 2 - 3 | 163 | 115 | ||
| GenBank mRNA | BC051549.1 | - | 2 - 3 | 115 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ras homolog gene family, member D |
Rhod |
AI326383, Arhd, Rho, RhoHP1, RhoM |
-4.3kb | Anti-sense | Chr. 19 - 4425457 - 4439432 on NCBI Build GRCm38 |
| Downstream | synaptotagmin XII | Syt12 | 1.9kb | Anti-sense | Chr. 19 - 4445908 - 4477143 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

A930001C03Rik
Genomic Map