Assay Details
| Gene Symbol: A930001C03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045989.1 | 3 - 4 | 191 | 115 | ||
| GenBank mRNA | BC051549.1 | - | 3 - 4 | 199 | 115 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | synaptotagmin XII | Syt12 | Anti-sense | Chr. 19 - 4445908 - 4477143 on NCBI Build GRCm38 | ||
| Downstream | RIKEN cDNA 2010003K11 gene | 2010003K11Rik | 49.4kb | Anti-sense | Chr. 19 - 4496788 - 4498583 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

A930001C03Rik
Genomic Map