Assay Details
| Gene Symbol: Slc3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009205.2 | NP_033231.2 | 7 - 8 | 1416 | 70 | |
| GenBank mRNA | AK052623.1 | - | 7 - 8 | 1387 | 70 | |
| AK132275.1 | - | 7 - 8 | 1388 | 70 | ||
| BC013441.1 | - | 7 - 8 | 1358 | 4222972 | 70 | |
| D88533.1 | - | 7 - 8 | 1376 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc3a1
Genomic Map