Assay Details
| Gene Symbol: Arih2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011790.4 | NP_035920.1 | 10 - 11 | 1244 | 75 | |
| GenBank mRNA | AF124664.1 | - | 9 - 10 | 1071 | 75 | |
| AJ130975.1 | - | 9 - 10 | 1103 | 75 | ||
| AK011094.1 | - | 10 - 11 | 1251 | 75 | ||
| AK045012.1 | - | 1227 | 75 | |||
| AK149936.1 | - | 10 - 11 | 1224 | 75 | ||
| AK167104.1 | - | 10 - 11 | 1201 | 75 | ||
| AK168055.1 | - | 10 - 11 | 1249 | 75 | ||
| BC051998.1 | - | 10 - 11 | 1244 | 6400287 | 75 | |
| BC052422.1 | - | 10 - 11 | 1244 | 6400287 | 75 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Arih2
Genomic Map