Assay Details
| Gene Symbol: Arih2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011790.4 | NP_035920.1 | 1 - 2 | 155 | 125 | |
| GenBank mRNA | AK011094.1 | - | 1 - 2 | 159 | 125 | |
| AK045012.1 | - | 138 | 125 | |||
| AK149936.1 | - | 1 - 2 | 138 | 125 | ||
| AK165028.1 | - | 1 - 2 | 93 | 125 | ||
| AK168055.1 | - | 1 - 2 | 163 | 125 | ||
| BC051998.1 | - | 1 - 2 | 155 | 6400287 | 125 | |
| BC052422.1 | - | 1 - 2 | 155 | 6400287 | 125 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK149936.1,BC051998.1,BC052422.1,NM_011790.4 transcripts.
This assay is in the 5'UTR region of the AK149936.1,BC051998.1,BC052422.1,NM_011790.4 transcripts.
Set Membership: |
|

Arih2
Genomic Map