Assay Details
| Gene Symbol: Arih2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011790.4 | NP_035920.1 | 7 - 8 | 957 | 108 | |
| GenBank mRNA | AF124664.1 | - | 6 - 7 | 784 | 108 | |
| AJ130975.1 | - | 6 - 7 | 816 | 108 | ||
| AK045012.1 | - | 940 | 108 | |||
| AK149936.1 | - | 7 - 8 | 937 | 108 | ||
| AK167104.1 | - | 7 - 8 | 914 | 108 | ||
| AK168055.1 | - | 7 - 8 | 962 | 108 | ||
| BC051998.1 | - | 7 - 8 | 957 | 6400287 | 108 | |
| BC052422.1 | - | 7 - 8 | 957 | 6400287 | 108 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Arih2
Genomic Map