Assay Details
| Gene Symbol: Eif2ak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011163.4 | NP_035293.1 | 13 - 14 | 1451 | 91 | |
| GenBank mRNA | AK028602.1 | - | 13 - 14 | 1451 | 91 | |
| BC016422.1 | - | 13 - 14 | 3964935 | 91 | ||
| M65029.1 | - | 13 - 14 | 1403 | 91 | ||
| M93567.1 | - | 13 - 14 | 1393 | 91 | ||
| S48937.1 | - | 3 - 4 | 431 | 91 | ||
| X62550.1 | - | 3 - 4 | 431 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eif2ak2
Genomic Map