Assay Details
| Gene Symbol: Lactb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145381.2 | NP_663356.1 | 4 - 5 | 654 | 138 | |
| GenBank mRNA | AK088932.1 | - | 4 - 5 | 667 | 138 | |
| AK090091.1 | - | 4 - 5 | 684 | 138 | ||
| AK143912.1 | - | 4 - 5 | 633 | 138 | ||
| BC004045.1 | - | 4 - 5 | 654 | 3496398 | 138 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lactb2
Genomic Map