Assay Details
| Gene Symbol: Rad54l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030730.2 | NP_109655.2 | 19 - 20 | 3512 | 60 | |
| GenBank mRNA | AJ132389.1 | - | 19 - 20 | 3413 | 60 | |
| AK143506.1 | - | 18 - 19 | 3128 | 60 | ||
| AK155151.1 | - | 19 - 20 | 3469 | 60 | ||
| BC133714.1 | - | 19 - 20 | 3361 | 40143565 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rad54l2
Genomic Map