Assay Details
| Gene Symbol: Iqcf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026645.3 | NP_080921.2 | 5 - 6 | 424 | 78 | |
| GenBank mRNA | AK016688.1 | - | 5 - 6 | 425 | 78 | |
| AK018835.1 | - | 5 - 6 | 427 | 78 | ||
| AK132739.1 | - | 5 - 6 | 429 | 78 | ||
| BC049549.1 | - | 5 - 6 | 425 | 6774389 | 78 | |
| DQ153247.1 | - | 5 - 6 | 430 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Iqcf3
Genomic Map