Assay Details
| Gene Symbol: Iqcf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026645.3 | NP_080921.2 | 7 - 8 | 652 | 65 | |
| GenBank mRNA | AK016688.1 | - | 7 - 8 | 653 | 65 | |
| AK018835.1 | - | 7 - 8 | 655 | 65 | ||
| AK132739.1 | - | 7 - 8 | 657 | 65 | ||
| BC049549.1 | - | 7 - 8 | 653 | 6774389 | 65 | |
| DQ153247.1 | - | 7 - 8 | 658 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P471746 | AK132739.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P471746 | BC049549.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P471746 | DQ153247.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P471746 | AK016688.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P471746 | AK018835.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P471746 | AK132739.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P471746 | BC049549.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P471746 | DQ153247.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P471746 | AK016688.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P471746 | AK018835.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 771966 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 784085 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925652 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 313831 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 349370 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 535571 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 372685 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 366627 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 702115 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 759560 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 78327 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105327 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145906 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P471746 | AK132739.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P471746 | BC049549.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P471746 | DQ153247.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P471746 | AK016688.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P471746 | AK018835.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P471746 | AK132739.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P471746 | BC049549.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P471746 | DQ153247.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P471746 | AK016688.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P471746 | AK018835.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26804019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26804013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26804009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26804012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26804006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26804018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26804021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26804004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26804028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26804024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26804026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26804022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26804015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26804016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26804000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26804020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26804003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803999 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26804010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26804005 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145906 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105327 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06915233 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02023070 | - | - |
| Whole Mouse Genome G4122A | A_51_P471746 | AK132739.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P471746 | BC049549.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P471746 | DQ153247.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P471746 | AK016688.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P471746 | AK018835.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P471746 | AK132739.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P471746 | BC049549.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P471746 | DQ153247.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P471746 | AK016688.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P471746 | AK018835.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113535 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131268 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06915233 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145906 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105327 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07752293 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07752298 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07752294 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07752299 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07752290 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07752292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26804022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804029 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07457492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08024199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803905 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07733496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07743015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803933 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07487952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07942627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803908 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07947174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07837902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07895664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804013 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803922 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07837904 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07767314 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803901 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07895407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07487949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803945 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07487947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07740463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07942634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08102526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804016 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07829276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26804021 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10596516 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10596513 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10596511 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10596515 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10596510 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10596514 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418056_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438990_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10596509 | - | - |
| Mouse Genome 430 2.0 Array | 1418056_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418056_at | - | - |
| Mouse Genome 430 2.0 Array | 1438990_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1438990_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68265_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68265_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900025052.mm.1 | NM_026645.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900025052.mm.1 | NM_026645.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900025052.mm.1 | TC0900003013.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1438990_PM_x_at | - | - |
| Mouse Expression Set 430 | 1418056_at | - | - |
| Mouse Expression Set 430 | 1438990_x_at | - | - |
| Mouse Expression Set 430 | 1418056_PM_at | - | - |
| Murine Genome U74v2 Set | 167646_f_at | - | - |
| Murine Genome U74v2 Set | 167646_PM_f_at | - | - |
| Murine Genome U74v2 Set | 165480_i_at | - | - |
| Murine Genome U74v2 Set | 165480_PM_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418056_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438990_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438990_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418056_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418056_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438990_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418056_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438990_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5570008 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005570008 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5570008 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl068265.1_222-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5570008 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2787448 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2787448 | - | - |

Iqcf3
Genomic Map