Assay Details
| Gene Symbol: A230046K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033375.2 | NP_001028547.2 | 15 - 16 | 1548 | 74 | |
| GenBank mRNA | AK038582.1 | - | 15 - 16 | 1506 | 74 | |
| AK043911.1 | - | 15 - 16 | 1519 | 74 | ||
| AK129269.1 | - | 10 - 11 | 1078 | 74 | ||
| AK145000.1 | - | 15 - 16 | 1530 | 74 | ||
| AK166233.1 | - | 15 - 16 | 1521 | 74 | ||
| BC020314.2 | - | 12 - 13 | 1183 | 74 | ||
| BC026467.1 | - | 4 - 5 | 509 | 74 | ||
| BC145215.1 | - | 15 - 16 | 1743 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

A230046K03Rik
Genomic Map