Assay Details
| Gene Symbol: A230046K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033375.2 | NP_001028547.2 | 18 - 19 | 1886 | 143 | |
| GenBank mRNA | AK038582.1 | - | 18 - 19 | 1844 | 143 | |
| AK043911.1 | - | 18 - 19 | 1857 | 143 | ||
| AK129269.1 | - | 13 - 14 | 1416 | 143 | ||
| AK145000.1 | - | 18 - 19 | 1868 | 143 | ||
| AK166233.1 | - | 18 - 19 | 1859 | 143 | ||
| BC020314.2 | - | 15 - 16 | 1521 | 143 | ||
| BC026467.1 | - | 7 - 8 | 847 | 143 | ||
| BC145215.1 | - | 18 - 19 | 2081 | 143 | ||
More Information
Updated Mapping Information
Set Membership: |
|

A230046K03Rik
Genomic Map