Assay Details
| Gene Symbol: A230046K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033375.2 | NP_001028547.2 | 19 - 20 | 2000 | 100 | |
| GenBank mRNA | AK038582.1 | - | 19 - 20 | 1958 | 100 | |
| AK043911.1 | - | 19 - 20 | 1971 | 100 | ||
| AK129269.1 | - | 14 - 15 | 1530 | 100 | ||
| AK166233.1 | - | 19 - 20 | 1973 | 100 | ||
| BC020314.2 | - | 16 - 17 | 1635 | 100 | ||
| BC026467.1 | - | 8 - 9 | 961 | 100 | ||
| BC145215.1 | - | 19 - 20 | 2195 | 100 | ||
More Information
Set Membership: |
|

A230046K03Rik
Genomic Map