Assay Details
| Gene Symbol: A230046K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033375.2 | NP_001028547.2 | 20 - 21 | 2113 | 82 | |
| GenBank mRNA | AK038582.1 | - | 20 - 21 | 2071 | 82 | |
| AK043911.1 | - | 20 - 21 | 2084 | 82 | ||
| AK129269.1 | - | 15 - 16 | 1643 | 82 | ||
| AK145000.1 | - | 20 - 21 | 2095 | 82 | ||
| AK166233.1 | - | 20 - 21 | 2086 | 82 | ||
| BC020314.2 | - | 17 - 18 | 1748 | 82 | ||
| BC026467.1 | - | 9 - 10 | 1074 | 82 | ||
| BC145215.1 | - | 20 - 21 | 2308 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

A230046K03Rik
Genomic Map