| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000007603.mm.1 |
NM_001033375.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007603.mm.1 |
NM_001033375.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007603.mm.1 |
TC1000001010.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000007606.mm.1 |
NM_001033375.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007606.mm.1 |
NM_001033375.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007606.mm.1 |
TC1000001010.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000007604.mm.1 |
NM_001033375.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007604.mm.1 |
NM_001033375.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007604.mm.1 |
TC1000001010.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000007605.mm.1 |
NM_001033375.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007605.mm.1 |
NM_001033375.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007605.mm.1 |
TC1000001010.mm.1 |
MTA TC |