Assay Details
| Gene Symbol: A230046K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033375.2 | NP_001028547.2 | 31 - 32 | 3457 | 113 | |
| GenBank mRNA | AK038582.1 | - | 31 - 32 | 3415 | 113 | |
| AK043911.1 | - | 31 - 32 | 3428 | 113 | ||
| AK089097.1 | - | 2 - 3 | 264 | 112 | ||
| AK129269.1 | - | 26 - 27 | 2987 | 113 | ||
| AK145000.1 | - | 31 - 32 | 3439 | 113 | ||
| AK161247.1 | - | 8 - 9 | 905 | 113 | ||
| AK166233.1 | - | 31 - 32 | 3430 | 113 | ||
| BC020314.2 | - | 28 - 29 | 3092 | 113 | ||
| BC026467.1 | - | 20 - 21 | 2418 | 113 | ||
| BC145215.1 | - | 31 - 32 | 3652 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

A230046K03Rik
Genomic Map