Assay Details
| Gene Symbol: A230046K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033375.2 | NP_001028547.2 | 10 - 11 | 887 | 113 | |
| GenBank mRNA | AK038582.1 | - | 10 - 11 | 845 | 113 | |
| AK043911.1 | - | 10 - 11 | 858 | 113 | ||
| AK129269.1 | - | 5 - 6 | 417 | 113 | ||
| AK145000.1 | - | 10 - 11 | 869 | 113 | ||
| AK166233.1 | - | 10 - 11 | 860 | 113 | ||
| BC020314.2 | - | 7 - 8 | 522 | 113 | ||
| BC145215.1 | - | 10 - 11 | 1082 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

A230046K03Rik
Genomic Map