Assay Details
| Gene Symbol: A230046K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033375.2 | NP_001028547.2 | 11 - 12 | 1011 | 88 | |
| GenBank mRNA | AK038582.1 | - | 11 - 12 | 969 | 88 | |
| AK043911.1 | - | 11 - 12 | 982 | 88 | ||
| AK129269.1 | - | 6 - 7 | 541 | 88 | ||
| AK145000.1 | - | 11 - 12 | 993 | 88 | ||
| AK166233.1 | - | 11 - 12 | 984 | 88 | ||
| BC020314.2 | - | 8 - 9 | 646 | 88 | ||
| BC145215.1 | - | 11 - 12 | 1206 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

A230046K03Rik
Genomic Map