Assay Details
| Gene Symbol: A230046K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033375.2 | NP_001028547.2 | 12 - 13 | 1144 | 153 | |
| GenBank mRNA | AK038582.1 | - | 12 - 13 | 1102 | 153 | |
| AK043911.1 | - | 12 - 13 | 1115 | 153 | ||
| AK129269.1 | - | 7 - 8 | 674 | 153 | ||
| AK145000.1 | - | 12 - 13 | 1126 | 153 | ||
| AK166233.1 | - | 12 - 13 | 1117 | 153 | ||
| BC020314.2 | - | 9 - 10 | 779 | 153 | ||
| BC026467.1 | - | 1 - 2 | 105 | 153 | ||
| BC145215.1 | - | 12 - 13 | 1339 | 153 | ||
More Information
Updated Mapping Information
Set Membership: |
|

A230046K03Rik
Genomic Map