Assay Details
| Gene Symbol: A230046K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033375.2 | NP_001028547.2 | 13 - 14 | 1291 | 73 | |
| GenBank mRNA | AK038582.1 | - | 13 - 14 | 1249 | 73 | |
| AK043911.1 | - | 13 - 14 | 1262 | 73 | ||
| AK129269.1 | - | 8 - 9 | 821 | 73 | ||
| AK145000.1 | - | 13 - 14 | 1273 | 73 | ||
| AK166233.1 | - | 13 - 14 | 1264 | 73 | ||
| BC020314.2 | - | 10 - 11 | 926 | 73 | ||
| BC026467.1 | - | 2 - 3 | 252 | 73 | ||
| BC145215.1 | - | 13 - 14 | 1486 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

A230046K03Rik
Genomic Map