Assay Details
| Gene Symbol: A230046K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033375.2 | NP_001028547.2 | 14 - 15 | 1418 | 114 | |
| GenBank mRNA | AK038582.1 | - | 14 - 15 | 1376 | 114 | |
| AK043911.1 | - | 14 - 15 | 1389 | 114 | ||
| AK129269.1 | - | 9 - 10 | 948 | 114 | ||
| AK145000.1 | - | 14 - 15 | 1400 | 114 | ||
| AK166233.1 | - | 14 - 15 | 1391 | 114 | ||
| BC020314.2 | - | 11 - 12 | 1053 | 114 | ||
| BC026467.1 | - | 3 - 4 | 379 | 114 | ||
| BC145215.1 | - | 14 - 15 | 1613 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

A230046K03Rik
Genomic Map