| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000019529.mm.1 |
NM_145220.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000019529.mm.1 |
NM_145220.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000019529.mm.1 |
TC1000002586.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000019534.mm.1 |
NM_145220.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000019534.mm.1 |
NM_145220.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000019534.mm.1 |
TC1000002586.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000019532.mm.1 |
NM_145220.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000019532.mm.1 |
NM_145220.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000019532.mm.1 |
TC1000002586.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000019527.mm.1 |
NM_145220.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000019527.mm.1 |
NM_145220.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000019527.mm.1 |
TC1000002586.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000019530.mm.1 |
NM_145220.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000019530.mm.1 |
NM_145220.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000019530.mm.1 |
TC1000002586.mm.1 |
MTA TC |