| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700024691.mm.1 |
NM_176917.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700024691.mm.1 |
NM_176917.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700024691.mm.1 |
TC1700002755.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700024695.mm.1 |
NM_176917.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700024695.mm.1 |
NM_176917.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700024695.mm.1 |
TC1700002755.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700024688.mm.1 |
NM_176917.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700024688.mm.1 |
NM_176917.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700024688.mm.1 |
TC1700002755.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700024687.mm.1 |
NM_176917.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700024687.mm.1 |
NM_176917.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700024687.mm.1 |
TC1700002755.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700024692.mm.1 |
NM_176917.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700024692.mm.1 |
NM_176917.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700024692.mm.1 |
TC1700002755.mm.1 |
MTA TC |