Assay Details
| Gene Symbol: Mettl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176917.4 | NP_795891.2 | 4 - 5 | 1395 | 75 | |
| GenBank mRNA | AK157268.1 | - | 4 - 5 | 1396 | 75 | |
| BC099943.1 | - | 3 - 4 | 1812 | 30020588 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mettl4
Genomic Map