Assay Details
| Gene Symbol: Mdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008618.3 | NP_032644.3 | 4 - 5 | 581 | 82 | |
| GenBank mRNA | AK002395.1 | - | 4 - 5 | 460 | 82 | |
| AK002404.1 | - | 4 - 5 | 440 | 81 | ||
| AK005237.1 | - | 4 - 5 | 476 | 82 | ||
| AK008270.1 | - | 4 - 5 | 453 | 82 | ||
| AK164785.1 | - | 4 - 5 | 462 | 82 | ||
| AK168545.1 | - | 4 - 5 | 419 | 82 | ||
| BC050940.1 | - | 4 - 5 | 447 | 6820601 | 82 | |
| M29462.1 | - | 4 - 5 | 460 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mdh1
Genomic Map