Assay Details
| Gene Symbol: Cabp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144532.2 | NP_653115.1 | 1 - 2 | 414 | 93 | |
| GenBank mRNA | AK171863.1 | - | 5 - 6 | 1024 | 93 | |
| AY039218.1 | - | 1 - 2 | 409 | 93 | ||
| BC029002.1 | - | 1 - 2 | 322 | 93 | ||
| BC049263.1 | - | 1 - 2 | 414 | 5366999 | 93 | |
| BC053330.1 | - | 1 - 2 | 369 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cabp4
Genomic Map