Assay Details
| Gene Symbol: Acy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF356878.1 | - | 2 - 3 | 168 | 72 | |
| AK002247.1 | - | 2 - 3 | 167 | 72 | ||
| AK143782.1 | - | 2 - 3 | 170 | 72 | ||
| AY169234.1 | - | 2 - 3 | 167 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF356878.1, AK002247.1, AK143782.1, AY169234.1 transcripts.
Set Membership: |
|

Acy3
Genomic Map