| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900000264.mm.1 |
NM_025529.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000264.mm.1 |
NM_025529.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000264.mm.1 |
TC1900000024.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900000267.mm.1 |
NM_025529.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000267.mm.1 |
NM_025529.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000267.mm.1 |
TC1900000024.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900000268.mm.1 |
NM_025529.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000268.mm.1 |
NM_025529.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000268.mm.1 |
TC1900000024.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900000270.mm.1 |
NM_025529.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000270.mm.1 |
NM_025529.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000270.mm.1 |
TC1900000024.mm.1 |
MTA TC |