Assay Details
| Gene Symbol: Socs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK033206.1 | - | 4 - 5 | 1206 | 71 | |
| BC027653.1 | - | 1 - 2 | 157 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Socs2| Gene Symbol: Socs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK033206.1 | - | 4 - 5 | 1206 | 71 | |
| BC027653.1 | - | 1 - 2 | 157 | 71 | ||
Set Membership: |
|