Assay Details
| Gene Symbol: Pja2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025309.1 | NP_001020480.1 | 4 - 5 | 1455 | 76 | |
| GenBank mRNA | AF493070.1 | - | 4 - 5 | 1512 | 76 | |
| AK122282.1 | - | 4 - 5 | 1414 | 76 | ||
| AK144586.1 | - | 4 - 5 | 1442 | 76 | ||
| AK164025.1 | - | 4 - 5 | 1460 | 76 | ||
| BC004742.1 | - | 1 - 2 | 299 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pja2
Genomic Map