Assay Details
| Gene Symbol: Pja2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025309.1 | NP_001020480.1 | 5 - 6 | 1636 | 67 | |
| GenBank mRNA | AF493070.1 | - | 5 - 6 | 1693 | 67 | |
| AK122282.1 | - | 5 - 6 | 1595 | 67 | ||
| AK144586.1 | - | 5 - 6 | 1623 | 67 | ||
| BC004742.1 | - | 2 - 3 | 480 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pja2
Genomic Map