Assay Details
| Gene Symbol: Gtf2f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133801.2 | NP_598562.1 | 2 - 3 | 224 | 107 | |
| GenBank mRNA | AK013002.1 | - | 2 - 3 | 238 | 107 | |
| AK078048.1 | - | 2 - 3 | 237 | 107 | ||
| AK168236.1 | - | 2 - 3 | 237 | 107 | ||
| AK168243.1 | - | 2 - 3 | 237 | 107 | ||
| BC003336.1 | - | 2 - 3 | 222 | 107 | ||
| BC023081.1 | - | 2 - 3 | 224 | 5355475 | 107 | |
| BC031123.1 | - | 2 - 3 | 354 | 5026448 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gtf2f1
Genomic Map