Assay Details
| Gene Symbol: Rab21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024454.1 | NP_077774.1 | 1 - 2 | 397 | 96 | |
| GenBank mRNA | AB232615.1 | - | 1 - 2 | 152 | 96 | |
| AK172891.1 | - | 1 - 2 | 397 | 96 | ||
| BC055042.1 | - | 1 - 2 | 397 | 6490069 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab21
Genomic Map