Assay Details
| Gene Symbol: Map2k3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008928.4 | NP_032954.1 | 9 - 10 | 934 | 79 | |
| GenBank mRNA | AK008141.1 | - | 9 - 10 | 937 | 79 | |
| AK011002.1 | - | 9 - 10 | 935 | 79 | ||
| AK150067.1 | - | 9 - 10 | 935 | 79 | ||
| AK159379.1 | - | 9 - 10 | 933 | 79 | ||
| AK166255.1 | - | 9 - 10 | 936 | 79 | ||
| AK166507.1 | - | 9 - 10 | 935 | 79 | ||
| AK167209.1 | - | 9 - 10 | 935 | 79 | ||
| AK167442.1 | - | 9 - 10 | 938 | 79 | ||
| AK168710.1 | - | 9 - 10 | 917 | 79 | ||
| BC007467.1 | - | 8 - 9 | 873 | 3497714 | 79 | |
| D87115.1 | - | 9 - 10 | 947 | 79 | ||
| X93150.1 | - | 7 - 8 | 669 | 79 | ||
More Information
Set Membership: |
|

Map2k3
Genomic Map