Assay Details
| Gene Symbol: Lclat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081071.2 | NP_001074540.1 | 4 - 5 | 704 | 96 | |
| NM_001177967.1 | NP_001171438.1 | 4 - 5 | 683 | 96 | ||
| NM_001177968.1 | NP_001171439.1 | 4 - 5 | 672 | 96 | ||
| GenBank mRNA | AK144580.1 | - | 4 - 5 | 647 | 96 | |
| BC147499.1 | - | 4 - 5 | 622 | 9056361 | 96 | |
| BC150884.1 | - | 4 - 5 | 611 | 9087795 | 96 | |
| BC158076.1 | - | 4 - 5 | 643 | 9088168 | 96 | |
| BC158079.1 | - | 4 - 5 | 643 | 9088171 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lclat1
Genomic Map