Assay Details
| Gene Symbol: Srd5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053188.2 | NP_444418.1 | 2 - 3 | 536 | 69 | |
| GenBank mRNA | AB049456.1 | - | 2 - 3 | 536 | 69 | |
| AK138946.1 | - | 2 - 3 | 477 | 69 | ||
| BC125510.1 | - | 2 - 3 | 520 | 40130025 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srd5a2
Genomic Map