Assay Details
| Gene Symbol: Arx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB006103.2 | - | 3 - 4 | 1163 | 70 | |
| BC052033.1 | - | 2 - 3 | 1157 | 5707995 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arx
Genomic Map