Assay Details
| Gene Symbol: 4930583I09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019827.1 | - | 3 - 4 | 397 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | down-regulated in hepatocellular carcinoma | Dreh | -64.6kb | Anti-sense | Chr. 17 - 64767111 - 64767931 on NCBI Build GRCm38 | |
| Downstream | transmembrane protein 232 | Tmem232 |
E130009J12Rik, Gm940, Tes13-L, Tes13-S |
421.7kb | Sense | Chr. 17 - 65255999 - 65541102 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930583I09Rik
Genomic Map