Assay Details
| Gene Symbol: 4732465J04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK169506.1 | - | 150 | 126 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nudix (nucleoside diphosphate linked moiety X)-type motif 4 | Nudt4 |
4933436C10Rik, DIPP2, DIPP2alpha, DIPP2beta, HDCMB47P |
-181.9kb | Anti-sense | Chr. 10 - 95547007 - 95564167 on NCBI Build GRCm38 |
| Downstream | early endosome antigen 1 | Eea1 |
A430109M19Rik, B230358H09Rik, ZFYVE2 |
167.0kb | Sense | Chr. 10 - 95940660 - 96045518 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4732465J04Rik
Genomic Map