Assay Details
| Gene Symbol: Khdrbs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133235.2 | NP_573498.1 | 1 - 2 | 199 | 103 | |
| GenBank mRNA | AK051807.1 | - | 1 - 2 | 230 | 103 | |
| AK051938.1 | - | 1 - 2 | 277 | 103 | ||
| AK078160.1 | - | 1 - 2 | 279 | 103 | ||
| BC066814.1 | - | 1 - 2 | 257 | 103 | ||
| BC132117.1 | - | 1 - 2 | 189 | 40130394 | 103 | |
| BC132119.1 | - | 1 - 2 | 189 | 40130396 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Khdrbs2
Genomic Map