Assay Details
| Gene Symbol: Khdrbs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133235.2 | NP_573498.1 | 3 - 4 | 454 | 62 | |
| GenBank mRNA | BC132117.1 | - | 3 - 4 | 444 | 40130394 | 62 |
| BC132119.1 | - | 3 - 4 | 444 | 40130396 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Khdrbs2
Genomic Map