Assay Details
| Gene Symbol: Gsta3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077353.2 | NP_001070821.1 | 2 - 3 | 171 | 56 | |
| GenBank mRNA | AI047326.1 | - | 2 - 3 | 149 | 56 | |
| BC147272.1 | - | 1 - 2 | 39 | 9056327 | 56 | |
| BC147273.1 | - | 1 - 2 | 39 | 9056339 | 56 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC147272.1, BC147273.1, NM_001077353.2 transcripts.
Set Membership: |
|

Gsta3
Genomic Map