Assay Details
| Gene Symbol: Yif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026553.4 | NP_080829.1 | 6 - 7 | 820 | 57 | |
| GenBank mRNA | AK002259.1 | - | 2 - 3 | 225 | 57 | |
| AK011071.1 | - | 6 - 7 | 815 | 57 | ||
| AK077329.1 | - | 6 - 7 | 806 | 57 | ||
| AK160409.1 | - | 6 - 7 | 773 | 57 | ||
| BC011117.1 | - | 6 - 7 | 762 | 4190535 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Yif1a
Genomic Map