Assay Details
| Gene Symbol: Zfp711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177747.3 | NP_808415.2 | 5 - 6 | 1317 | 75 | |
| GenBank mRNA | CN525862.1 | - | 3 - 4 | 427 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp711
Genomic Map