Assay Details
| Gene Symbol: Eya1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252192.1 | NP_001239121.1 | 12 - 13 | 1673 | 101 | |
| NM_010164.2 | NP_034294.2 | 13 - 14 | 1789 | 101 | ||
| GenBank mRNA | AK042412.1 | - | 12 - 13 | 1596 | 101 | |
| AK153788.1 | - | 13 - 14 | 1795 | 101 | ||
| AK162111.1 | - | 5 - 6 | 530 | 101 | ||
| BC060260.1 | - | 13 - 14 | 1789 | 6848408 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eya1
Genomic Map