Assay Details
| Gene Symbol: Sgk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291152.1 | NP_001278081.1 | 10 - 11 | 957 | 58 | |
| NM_001291154.1 | NP_001278083.1 | 10 - 11 | 1589 | 58 | ||
| NM_013731.3 | NP_038759.1 | 11 - 12 | 1075 | 58 | ||
| GenBank mRNA | AF169033.1 | - | 11 - 12 | 1010 | 58 | |
| AK050009.1 | - | 10 - 11 | 1570 | 58 | ||
| AK136559.1 | - | 11 - 12 | 1027 | 58 | ||
| BC026549.1 | - | 9 - 10 | 702 | 4973908 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgk2
Genomic Map