Assay Details
| Gene Symbol: Sgk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291152.1 | NP_001278081.1 | 1 - 2 | 246 | 91 | |
| NM_001291154.1 | NP_001278083.1 | 2 - 3 | 962 | 91 | ||
| NM_013731.3 | NP_038759.1 | 2 - 3 | 361 | 91 | ||
| GenBank mRNA | AF169033.1 | - | 2 - 3 | 296 | 91 | |
| AK050009.1 | - | 2 - 3 | 943 | 91 | ||
| AK136559.1 | - | 2 - 3 | 313 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF169033.1, AK050009.1, AK136559.1, NM_001291152.1, NM_001291154.1, NM_013731.3 transcripts.
Set Membership: |
|

Sgk2
Genomic Map