Assay Details
| Gene Symbol: Sgk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291154.1 | NP_001278083.1 | 6 - 7 | 1221 | 86 | |
| NM_013731.3 | NP_038759.1 | 6 - 7 | 620 | 86 | ||
| GenBank mRNA | AF169033.1 | - | 6 - 7 | 555 | 86 | |
| AK050009.1 | - | 6 - 7 | 1202 | 86 | ||
| AK136559.1 | - | 6 - 7 | 572 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sgk2
Genomic Map