Assay Details
| Gene Symbol: Cdc25a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007658.3 | NP_031684.3 | 11 - 12 | 1477 | 118 | |
| GenBank mRNA | AK036499.1 | - | 1 - 2 | 929 | 118 | |
| AK077609.1 | - | 11 - 12 | 1095 | 118 | ||
| AK077991.1 | - | 11 - 12 | 1480 | 117 | ||
| AK142334.1 | - | 10 - 11 | 1752 | 118 | ||
| BC046296.1 | - | 11 - 12 | 1096 | 6486343 | 118 | |
| U27323.1 | - | 11 - 12 | 1066 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdc25a
Genomic Map